missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
LaMotte™ DPD Free Chlorine Reagent Refills
The liquid alternative to DPD tablet refills
$31.94 - $39.50
Specifications
For Use With (Application) | Measurement of total chlorine, provides quicker results and clearer colors than tablets |
---|---|
For Use With (Equipment) | LaMotte™ colorimeters and comparators |
Quantity | Each |
Alert:
Keep out of reach of children. May irritate eyes and skin. Harmful if swallowed.
Catalog Number | Mfr. No. | Type | Volume (English) | Volume (Metric) | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Volume (English) | Volume (Metric) | Price | Quantity | ||||
22-278-647
|
LaMotte™
P6743CNG |
DPD 3 Liquid Reagent | 1 oz. | 30 mL |
N/A
|
|
||||
22-278-643
|
LaMotte™
P6740CNG |
DPD 1A Liquid Reagent | 1 oz. | 30 mL |
N/A
|
|
||||
22-278-645
|
LaMotte™
P6741CNG |
DPD 1B Liquid Reagent | 1 oz. | 30 mL |
N/A
|
|
||||
Description
- DPD £1A and DPD £1B test Free Available Chlorine (Cl)
- DPD £3 is added to the reacted sample to measure Total Cl
- Comes in 30mL and 60mL sizes
DPD = N, N-Diethyl-1, 4-phenylene-diamine sulfate
Use with existing LaMotte chlorine comparators or colorimeters. For each test, all three reagents are added to the same sample. Ideal for testing drinking water, food and beverage, pool and spa, industrial water boiling and cooling, laundry and sanitation, water and wastewater.
Specifications
Measurement of total chlorine, provides quicker results and clearer colors than tablets | |
LaMotte™ colorimeters and comparators | |
Each |