Learn More
Dräger™ Civil Defense Simultest Kit: Test Sets
For use with Civil Defense Simultest Kit (Cat. No. 19-038-865)
Supplier: Dräger™ 8103140
Alert:
No other type of tube should be substituted for use with these kits.
Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.
Description
For Chemical Warfare Agent (CWA) detection. test sets detect eight chemical agents, including nerve, blood, lung, nose, and throat irritants. Include five colorimetric DraegerTubes™, the same field-proven tube chemistry used by the North Atlantic Treaty Organization (NATO) in single detector tubes. Each test set provides on-the-spot results in less than five minutes.
CDS Test Set I detector tubes test for hydrocyanic acid, phosgene, organic arsenic compounds and arsine, organic basic nitrogen compounds, and thioether
CDS Test Set V detector tube test for phosphoric acid esters, phosgene, cyanogen chloride, chlorine, and thioether
Specifications
CDS Test Set I | |
Gas detection, no calibration | |
Includes colorimetric detector tubes for hydrocyanic acid, phosgene, organic arsenic compounds, arsine, organic basic nitrogen compounds, and thioether |