Learn More
Dräger™ Civil Defense Simultest Kit: Test Sets
For use with Civil Defense Simultest Kit (Cat. No. 19-038-865)
$586.34 - $586.34
Specifications
For Use With (Application) | Gas detection, no calibration |
---|
Alert:
No other type of tube should be substituted for use with these kits.
Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.
Catalog Number | Mfr. No. | Type | Includes | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Includes | Price | Quantity | ||||
19-046-635
|
Dräger™
8103140 |
CDS Test Set I | Includes colorimetric detector tubes for hydrocyanic acid, phosgene, organic arsenic compounds, arsine, organic basic nitrogen compounds, and thioether |
Pack of 5 Each for $586.34
|
|
||||
19-046-636
|
Dräger™
8103200 |
CDS Test Set V | Includes colorimetric detector tubes for phosphoric acid esters, phosgene, cyanogen chloride, chlorine, and thioether |
Pack of 5 Each for $586.34
|
|
||||
Description
For Chemical Warfare Agent (CWA) detection. test sets detect eight chemical agents, including nerve, blood, lung, nose, and throat irritants. Include five colorimetric DraegerTubes™, the same field-proven tube chemistry used by the North Atlantic Treaty Organization (NATO) in single detector tubes. Each test set provides on-the-spot results in less than five minutes.
CDS Test Set I detector tubes test for hydrocyanic acid, phosgene, organic arsenic compounds and arsine, organic basic nitrogen compounds, and thioether
CDS Test Set V detector tube test for phosphoric acid esters, phosgene, cyanogen chloride, chlorine, and thioether
Specifications
Gas detection, no calibration |