Dräger™ Short-Term Detector Tubes - De Tube Hydrochloric Acid
For making spot measurments for various gases and vapors
Supplier: Dräger™ CH29501
Description
- Glass vial
- Calibrated 100mL sample of air drawn through tube with Drager Accuro™ bellows pump
Specifications
Hydrochloric Acid 1/a | |
1 to 10ppm |