missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DR Instruments Iris Scissors, 4.5 in., Stainless Steel

Iris Scissors feature precision ground blades for effortless cutting. Ideal for delicate dissections.

$4.79 - $5.38

Specifications

Length (English) 4.5 in.
Length (Metric) 11.4 cm
Material Stainless Steel
For Use With (Application) Dissection
View More Specs

Alert:

California Proposition 65 WARNING: Cancer and Reproductive Harm


Products 2
Catalog Number Mfr. No. Tip Style Price Quantity  
Catalog Number Mfr. No. Tip Style Price Quantity  
S07897
View Documents
DR Instruments
10
Curved Fine Points
Each for $5.38
Case of 25 Each for $134.50
 
S08088
View Documents
DR Instruments
9
Fine Point
Each for $4.79
 
Description

Description

  • For fine dissection work
  • Surgical stainless steel construction
  • Available in sand finish or mirror polish finish
  • Length: 11.4 cm (4.5 in.)
Specifications

Specifications

4.5 in.
11.4 cm
Stainless Steel
Dissection
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'