missing translation for 'onlineSavingsMsg'
Learn More

DR Instruments Iris Scissors, 4.5 in., Stainless Steel

Iris Scissors feature precision ground blades for effortless cutting. Ideal for delicate dissections.

Supplier:  DR Instruments 9

 View more versions of this product

Catalog No. S08088


Only null left
Explore available promotions

Alert:

California Proposition 65 WARNING: Cancer and Reproductive Harm


Description

Description

  • For fine dissection work
  • Surgical stainless steel construction
  • Available in sand finish or mirror polish finish
  • Length: 11.4 cm (4.5 in.)
Specifications

Specifications

4.5 in.
Stainless Steel
Fine Point
11.4 cm
Dissection
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions