Fisherbrand™ Drosophila Products, BuzzPlugs™ Vial and Bottle Closures
Nonshedding, dense weave of nontoxic cellulose acetate minimizes evaporation, keeps food fresh longer
Supplier:  Fisherbrand™ 99102
 View more versions of this product
Catalog No. AS271
Explore available promotions
Description
Description
Nonshedding, dense weave of nontoxic cellulose acetate minimizes evaporation, keeps food fresh longer
  • Superior barrier to mite infestation
  • Exact fit with narrow 0.9 in. (25mm) or wide 1.1 in. (28.5mm) dia. plastic shell vials or standard (≈24mm dia.) glass vials or plastic stock bottles
  • Stretches yet maintains tight fit after repeated removal and closing
  • Flat surface accepts marking pen
Specifications
Specifications
Superior barrier
BuzzPlugs Bottle Closure
Cellulose Acetate
Minimizes evaporation, keeps food fresh longer
1000 Case
Standard (2.4cm dia.) glass vials