Fisherbrand™ Drosophila Products, BuzzPlugs™ Vial and Bottle Closures
Nonshedding, dense weave of nontoxic cellulose acetate minimizes evaporation, keeps food fresh longer
$344.25 - $827.56
Specifications
Material Cellulose Acetate
Type BuzzPlugs Bottle Closure
For Use With (Application) Minimizes evaporation, keeps food fresh longer
Includes Superior barrier
Quantity 1000 Case
View More Specs
Products 4
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity  
AS273
View Documents
N/A 2.5cm narrow plastic shell vials
Case of 1000 Each for $382.90
AS275
View Documents
N/A 2.8cm wide plastic shell vials
Case of 1000 Each for $427.59
AS271
View Documents
Fisherbrand™
99102
Standard (2.4cm dia.) glass vials
Case of 1000 Each for $344.25
AS277
View Documents
Fisherbrand™
20067
Standard (2.4cm dia.) plastic stock bottles
Case of 1000 Each for $827.56
Description
Description
Nonshedding, dense weave of nontoxic cellulose acetate minimizes evaporation, keeps food fresh longer
  • Superior barrier to mite infestation
  • Exact fit with narrow 0.9 in. (25mm) or wide 1.1 in. (28.5mm) dia. plastic shell vials or standard (≈24mm dia.) glass vials or plastic stock bottles
  • Stretches yet maintains tight fit after repeated removal and closing
  • Flat surface accepts marking pen
Specifications
Specifications
Cellulose Acetate
Minimizes evaporation, keeps food fresh longer
1000 Case
BuzzPlugs Bottle Closure
Superior barrier
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'