Excelta™ Duckbill Point Tweezers
Duckbill Point Tweezers used for handling labels and SMD components.
Supplier:  Excelta™ 2ASA
 View more versions of this product
Catalog No. 17456042
Explore available promotions
Description
Description
  • Anti-magnetic stainless steel or Carbon Steel or Magnetic Stainless Steel
Specifications
Specifications
Tapered, Flat Square Duckbilled
Anti-magnetic Stainless Steel
Tweezers
For handling labels and SMD components
0.038 cm
0.25 in.
High-precision
2.8 in.
71 mm
1 Ea.
0.015 in.
High
0.63 cm
Product Suggestions
Product Suggestions