Excelta™ Duckbill Point Tweezers
Duckbill Point Tweezers used for handling labels and SMD components.
$22.78 - $290.72
Specifications
Product Type | Tweezers |
---|---|
Quantity (No. per Case) | 1 Ea. |
For Use With (Application) | For handling labels and SMD components |
Style | High-precision |
Catalog Number | Mfr. No. | Length (English) | Material | Tip Style | Precision | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (English) | Material | Tip Style | Precision | Price | Quantity | |||
17467125
|
Excelta™
2ASAPI |
4.75 in. | Anti-magnetic Stainless Steel | Tapered, Flat Round Duckbilled | Fine |
Each for $113.72
|
|
|||
17447111
|
Excelta™
FSA |
4.75 in. | Anti-magnetic Stainless Steel | Tapered, Flat Square Duckbilled | High |
Each for $242.16
|
|
|||
17447112
|
Excelta™
FSAPI |
4.75 in. | Anti-magnetic Stainless Steel | Tapered, Flat Square Duckbilled | Fine |
Each for $93.59
|
|
|||
18100818
|
Excelta™
2ASSE |
4.75 in. | Stainless Steel | Tapered, Flat Round Duckbilled, Economy | High |
Each for $24.28
|
|
|||
17467130
|
Excelta™
2ABSASE |
4.5 in. | Anti-magnetic Stainless Steel | Curved, Flat Round, Duckbilled | General |
Each for $22.78
|
|
|||
18100814
|
Excelta™
XLT2A |
4.75 in. | Carbon Steel | Tapered, Flat Round Duckbilled | High |
Each for $206.84
|
|
|||
17456104
|
Excelta™
27SA |
4.25 in. | Anti-magnetic Stainless Steel | Flat | Flat tip |
Each for $263.34
|
|
|||
17467129
|
Excelta™
2ABSAPI |
4.5 in. | Anti-magnetic Stainless Steel | Curved, Flat Round, Duckbilled | Fine |
Each for $115.69
|
|
|||
18100817
|
Excelta™
2AS |
4.75 in. | Stainless Steel | Tapered, Flat Round Duckbilled | High |
Each for $187.97
|
|
|||
17467127
|
Excelta™
2ASASE |
4.75 in. | Anti-magnetic Stainless Steel | Tapered, Flat Round Duckbilled | General |
Each for $24.28
|
|
|||
17456053
|
Excelta™
25SA |
4.75 in. | Anti-magnetic Stainless Steel | Straight, Large Flat Round Duckbilled | High |
Each for $290.72
|
|
|||
17467128
|
Excelta™
2ABN |
4.5 in. | Anti-magnetic Nickel | Curved, Flat Round, Duckbilled | High |
Each for $217.88
|
|
|||
17456042
|
Excelta™
2ASA |
2.8 in. | Anti-magnetic Stainless Steel | Tapered, Flat Square Duckbilled | High |
Each for $167.88
|
|
|||
Description
- Anti-magnetic stainless steel or Carbon Steel or Magnetic Stainless Steel
Specifications
Tweezers | |
1 Ea. | |
For handling labels and SMD components | |
High-precision |