Brady™ Duct Markers: RELIEF AIR
Differentiate between different air systems. Brady™ Duct Markers are made of B-946 self-sticking vinyl.
Supplier:  Brady™ 98487
 View more versions of this product
Catalog No. 19-112-030
Explore available promotions
Description
Description
  • Color-coded to differentiate between different air systems
  • With detachable arrows which can be applied in any direction
  • Made of Pressure Sensitive Vinyl (B-964)
Specifications
Specifications
Abrasion and chemical resistant
White, Blue
Vinyl (B-946)
RELIEF AIR
Acrylic, Permanent, Cold-temperature
Die-cut
Duct Marker
13.25 x 2.25 in. (W x H)