missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DuPont™ Tyvek™ IsoClean™ 270 Series Sterile Frocks
Clean-processed and sterile frocks provide an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash.
$601.49 - $729.91
Specifications
Product Type | Controlled Environments Frock |
---|---|
Material | Tyvek |
Color | White |
Disposable | Single-use |
Sterility | Sterile |
Alert:
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Catalog Number | Mfr. No. | Size | Length (English) | Length (Metric) | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Length (English) | Length (Metric) | Price | Quantity | |||
19-161-156
|
DuPont™
IC270BWHSM0030CS |
Small | 39 in. | 99 cm |
Case of 30 Each for $601.49
|
|
|||
19-088-1127B
|
DuPont™
IC270BWHMD0030CS |
Medium | 40 in. | 101.6 cm |
Case of 30 Each for $601.49
|
|
|||
19-088-1127C
|
DuPont™
IC270BWHLG0030CS |
Large | 41 in. | 104.14 cm |
Case of 30 Each for $601.49
|
|
|||
19-088-1127D
|
DuPont™
IC270BWHXL0030CS |
X-Large | 43 in. | 109.2 cm |
Case of 30 Each for $601.49
|
|
|||
19-088-1127E
|
DuPont™
IC270BWH2X0030CS |
2X-Large | 44 in. | 111.76 cm |
Case of 30 Each for $637.62
|
|
|||
19-088-1127F
|
DuPont™
IC270BWH3X0030CS |
3X-Large | 45 in. | 114.3 cm |
Case of 30 Each for $675.86
|
|
|||
17-990-512
|
DuPont™
IC270BWH4X0030CS |
4X-Large | 46 in. | 116.8 cm |
Case of 30 Each for $729.91
|
|
|||
Description
- Garments are gamma sterilized to SAL of 10-6
- Body-length jacket design with front snap closures for easy donning and doffing
- Made of Tyvek™, a spunbonded olefin fabric that is lightweight and flexible
- Unique, patented flashspinning process creates an barrier to dry particles, microorganisms, and nonhazardous liquids
- Garments are constructed with the smooth side out to reduce particulate entrapment and bound seams reduce lint
- ISO Class 5 (formerly Class 100) compatible
- Processed in a certified ISO Class 4 (Class 10) cleanroom, antistatic treated and packaged in durable, antistatic bags
- Generous sizing permits freedom of movement
- Plastic snaps won't rust or flake during processing
- Bound seams are covered with garment fabric to reduce the potential for particle penetration
- Elastic opening for tighter fit at wrist
- Bound neck for lower particle shedding, adjustable snap at neck for better fit
- Full traceability
Specifications
Controlled Environments Frock | |
White | |
Sterile | |
Elastic | |
None | |
Long Sleeve | |
Class 100 (ISO 5) |
Tyvek | |
Single-use | |
Snaps | |
Bound | |
IsoClean™ 270 Series | |
Clean-processed, Individually Packaged |