Learn More
DuPont™ Tychem™ CPF 3 Combo Suit
Supplier:  DuPont™ C3750TTN3X000600
 View more versions of this product
Catalog No. 19-088-655
Only null left
Explore available promotions
Description
Description
Tychem 5000 fabric is composed of a multi-layer film barrier laminated to a durable 2.0 oz/yd2 polypropylene substrate. Tychem 5000 fabric is strong and durable for rigorous activities and rugged situations involving liquid splash and provides barrier to a broad range of chemicals. Typical Applications: chemical handling, petro-chemical, hazardous materials/waste clean-up, fire departments, industrial hazmat teams, utilities, and domestic preparedness. Commonly used in domestic preparedness for situations where the potential to exposure to chemicals exist.Jacket and bib overall combination ensemble. Components also sold separatelyComes with a jam fit cuff and glove insert for user supplied gloves
Specifications
Specifications
ASTM D751, ASTM D1117, ASTM D3787, ASTM D751, ASTM D2582, ASTM D751, 16CFR1610
Chemical handling, petro-chemical, hazardous materials/waste clean-up, fire departments, industrial hazmat teams, utilities, and domestic preparedness
Multi-layer Barrier Laminate Film
Open
Elastic
Single-use
189 to 195 cm
31.5 in.
76.2 cm
Taped
120 to 129 cm
Tan
Attached
3X-Large
Front Zipper with Double Storm Flap
Adjustable webbing, straps with closure; Multi-layer film barrier laminated
6.2 to 6.4 ft.
30 in.
36.7 in.
93.2 cm
30.25 in.
18 mil
Product Suggestions
Product Suggestions