missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DuPont™ Tychem™ CPF 3 Combo Suit
$830.58 - $933.20
Specifications
Certifications/Compliance | ASTM D751, ASTM D1117, ASTM D3787, ASTM D751, ASTM D2582, ASTM D751, 16CFR1610 |
---|---|
Color | Tan |
For Use With (Application) | Chemical handling, petro-chemical, hazardous materials/waste clean-up, fire departments, industrial hazmat teams, utilities, and domestic preparedness |
Hood Type | Attached |
Ankle Style | Open |
Catalog Number | Mfr. No. | Size | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | ||||||
19-088-651
|
DuPont™
C3750TTNSM000600 |
Small |
Case of 6 Each for $830.58
|
|
||||||
19-088-653
|
DuPont™
C3750TTNLG000600 |
Large |
Case of 6 Each for $830.58
|
|
||||||
19-088-652
|
DuPont™
C3750TTNMD000600 |
Medium |
Case of 6 Each for $830.58
|
|
||||||
19-088-654
|
DuPont™
C3750TTN2X000600 |
2X-Large |
Case of 6 Each for $880.35
|
|
||||||
19-088-655
|
DuPont™
C3750TTN3X000600 |
3X-Large |
Case of 6 Each for $933.20
|
|
||||||
Description
Tychem 5000 fabric is composed of a multi-layer film barrier laminated to a durable 2.0 oz/yd2 polypropylene substrate. Tychem 5000 fabric is strong and durable for rigorous activities and rugged situations involving liquid splash and provides barrier to a broad range of chemicals. Typical Applications: chemical handling, petro-chemical, hazardous materials/waste clean-up, fire departments, industrial hazmat teams, utilities, and domestic preparedness. Commonly used in domestic preparedness for situations where the potential to exposure to chemicals exist.Jacket and bib overall combination ensemble. Components also sold separatelyComes with a jam fit cuff and glove insert for user supplied glovesSpecifications
ASTM D751, ASTM D1117, ASTM D3787, ASTM D751, ASTM D2582, ASTM D751, 16CFR1610 | |
Chemical handling, petro-chemical, hazardous materials/waste clean-up, fire departments, industrial hazmat teams, utilities, and domestic preparedness | |
Open | |
Elastic | |
Single-use | |
18 mil |
Tan | |
Attached | |
Front Zipper with Double Storm Flap | |
Adjustable webbing, straps with closure; Multi-layer film barrier laminated | |
Taped |