DuPont™ Tychem™ 2000 Series 122 Coveralls
Flexible and durable light liquid splash protection
Supplier:  DuPont™ QC122SYL2X001200
 View more versions of this product
Catalog No. 29-635-3359
Explore available promotions
Description
Description
Tychem QC is a polyethylene-coated Tyvek™ fabric that provides at least 30 minutes of protection against 42 chemical challenges.
  • Attached hood with elastic around face opening
  • Elastic opening for tighter fit at wrist
  • Integrated socks composed of garment material
  • Yellow for high visibility
  • 10mil thickness
QC 122B
  • Tightly sewn seam is covered with garment fabric to reinforce seam and reduce potential for particle penetration
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
QC 122S
  • Multiple interlocking threads are sewn around the raw edges of garment material to create a strong, stress-resistant seam
  • QC 122T
    • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing
    • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
    Compliance
    Meets ASTM™ D1777, ASTM D3776, ASTM D3786, ASTM D5034 standards.
    Specifications
    Specifications
    2X-Large
    Yellow
    Elastic
    Attached
    Serged
    30 in.
    10 mil
    38.25 in.
    28.75 in.
    1.82 to 1.95 m
    Polyethylene-coated Tyvek
    Single-use
    Elastic
    Front Zipper
    One piece construction
    112.39 to 121.28 cm
    Petroleum refining, pulp and Paper manufacturing, food processing, chemical processing and pharmaceutical manufacturing
    97.15 cm
    6 to 6.4 ft.