missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DuPont™ Tychem™ 2000 Series 122 Coveralls

Flexible and durable light liquid splash protection

$193.44 - $361.39

Specifications

Color Yellow
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Hood Type Attached
View More Specs

Products 22
Catalog Number Mfr. No. Size Seam Type Price Quantity  
Catalog Number Mfr. No. Size Seam Type Price Quantity  
29-635-5554
missing translation for 'viewDocuments'
DuPont™
QC122BYLMD001200
Medium Bound N/A
 
19-014-971
missing translation for 'viewDocuments'
DuPont™
QC122BYLLG001200
Large Bound N/A
 
19-014-972
missing translation for 'viewDocuments'
DuPont™
QC122BYLXL001200
X-Large Bound N/A
 
19-085-218
missing translation for 'viewDocuments'
DuPont™
QC122BYL2X001200
2X-Large Bound N/A
 
19-085-219
missing translation for 'viewDocuments'
DuPont™
QC122BYL3X001200
3X-Large Bound N/A
 
19085220
missing translation for 'viewDocuments'
DuPont™
QC122BYL4X001200
4X-Large Bound N/A
 
19-141-760
missing translation for 'viewDocuments'
DuPont™
QC122BYL5X001200
5X-Large Bound N/A
 
19-085-222
missing translation for 'viewDocuments'
DuPont™
QC122BYL6X001200
6X-Large Bound N/A
 
29-635-3357
missing translation for 'viewDocuments'
DuPont™
QC122SYLMD001200
Medium Serged N/A
 
19-812-697
missing translation for 'viewDocuments'
DuPont™
QC122SYLLG001200
Large Serged N/A
 
29-635-3358
missing translation for 'viewDocuments'
DuPont™
QC122SYLXL001200
X-Large Serged N/A
 
29-635-3359
missing translation for 'viewDocuments'
DuPont™
QC122SYL2X001200
2X-Large Serged N/A
 
29-635-3360
missing translation for 'viewDocuments'
DuPont™
QC122SYL3X001200
3X-Large Serged N/A
 
19088326D
missing translation for 'viewDocuments'
DuPont™
QC122SYL4X001200
4X-Large Serged N/A
 
19-162-754
missing translation for 'viewDocuments'
DuPont™
QC122TYLMD000400
Medium Taped N/A
 
19-162-753
missing translation for 'viewDocuments'
DuPont™
QC122TYLLG000400
Large Taped N/A
 
19-162-755
missing translation for 'viewDocuments'
DuPont™
QC122TYLXL000400
X-Large Taped N/A
 
19-162-748
missing translation for 'viewDocuments'
DuPont™
QC122TYL2X000400
2X-Large Taped N/A
 
19-162-749
missing translation for 'viewDocuments'
DuPont™
QC122TYL3X000400
3X-Large Taped N/A
 
19-162-750
missing translation for 'viewDocuments'
DuPont™
QC122TYL4X000400
4X-Large Taped N/A
 
19-162-751
missing translation for 'viewDocuments'
DuPont™
QC122TYL5X000400
5X-Large Taped N/A
 
19-162-752
missing translation for 'viewDocuments'
DuPont™
QC122TYL6X000400
6X-Large Taped N/A
 
Description

Description

Tychem QC is a polyethylene-coated Tyvek™ fabric that provides at least 30 minutes of protection against 42 chemical challenges.

  • Attached hood with elastic around face opening
  • Elastic opening for tighter fit at wrist
  • Integrated socks composed of garment material
  • Yellow for high visibility
  • 10mil thickness

QC 122B

  • Tightly sewn seam is covered with garment fabric to reinforce seam and reduce potential for particle penetration
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
QC 122S
  • Multiple interlocking threads are sewn around the raw edges of garment material to create a strong, stress-resistant seam
  • QC 122T
    • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing
    • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper

    Compliance

    Meets ASTM™ D1777, ASTM D3776, ASTM D3786, ASTM D5034 standards.

    Specifications

    Specifications

    Yellow
    Elastic
    Attached
    One piece construction
    Single-use
    Elastic
    Front Zipper
    10 mil
    Videos
    SDS
    Documents

    Documents