missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DuPont™ Tychem™ 4000 Series 127 Coveralls

Ideal for chemical mixing, remediation, emergency medical response, paint spraying, and radioactive environments

$336.33 - $408.19

Specifications

Material Tychem
Color White
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
View More Specs

Alert:

Garments made of Tyvek™ should not be used around heat, flames, sparks or in potentially flammable or explosive environments.


Products 6
Catalog Number Mfr. No. Closure Type Seam Type Size Height (English) Price Quantity  
Catalog Number Mfr. No. Closure Type Seam Type Size Height (English) Price Quantity  
19-088-2005B
View Documents
DuPont™
SL127TWH3X000600
Front Zipper Taped 3X-Large 6.2 to 6.4 ft. N/A
 
19-087-152
View Documents
DuPont™
SL127TWH2X000600
Front Zipper Taped 2X-Large 6 to 6.4 ft. N/A
 
19-088-2005A
View Documents
DuPont™
SL127TWHXL000600
Front Zipper Taped X-Large 5.8 to 6.2 ft. N/A
 
19-088-2132
View Documents
DuPont™
SL127TWHMD000600
Front Zipper Taped Medium 5.3 to 5.7 ft. N/A
 
19-088-2131
View Documents
DuPont™
SL127TWHLG000600
Front Zipper Taped Large 5.5 to 5.9 ft. N/A
 
19-088-2134
View Documents
DuPont™
SL127TWH4X000600
Front Zipper Taped 4X-Large 6.4 to 6.7 ft. N/A
 
Description

Description

Lightweight, comfortable garment designed for easy wear. Excellent for waste management, PCB cleanup, hazardous response, and nuclear environments.

  • DuPont™Tyvek™ fabric laminated with Dow™ Saranex™-23P film
  • Attached hood
  • Elasticized face, wrists, and ankles
  • Zipper front with storm flap
  • Bound-seam models are tightly sewn with a reinforced outer binding to increase strength and particle barrier
  • Taped-seam models are sewn and taped to provide chemical resistance against heavy liquid splashes and tough seam stress
  • Compliance

    Meets ASTM™ D3776, ASTM D5034, ASTM D1777, ASTM F739, ASTM D3786; 16 CFR 1610 Standard for Flammability of Clothing Textiles; MIL-STD-282 requirements.

Specifications

Specifications

Tychem
Elastic
Hooded
Taped
12 mil
0.22 lb.
White
Elastic
Front Zipper
ASTM D3776, ASTM D5034, ASTM D1777, ASTM F739, ASTM D3786, 16 CFR 1610, MIL-STD-282
Chemical mixing, hazardous remediation, emergency medical response, paint spraying and nuclear environments
0.1 kg
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'