DuPont™ Tychem™ TK Encapsulated Level A Suit (Rear Entry)
Encapsulated Level A garment design provides highest level of protection from liquid splash and vapor/gas exposures for both the wearer and respiratory equipment
Supplier:  DuPont™ TK613TLY3X000100
 View more versions of this product
Catalog No. 19-067-046
Explore available promotions
Description
Description
 Tychem™ TK exhibits excellent chemical barrier properties and offers an extremely durable fabric that is puncture- and tear-resistant. It shows at least 30 minutes of barrier protection to 322 challenge chemicals with no observed breakthrough. Fabris is specifically developed for protection against toxic, corrosive gases, liquids and solid chemicals. 
  • High-performance chemical barrier fabric is suited for industrial, HazMat and domestic preparedness applications
  • Class 2 garments are designed for situations where there is an immediate danger to life and health (IDLH) and must be worn with self-contained breathing apparatus (SCBA)
  • Double taped seams provide strong chemical resistance against heavy liquid splashes
  • Expanded faceshield provides wide 220° viewing angle
  • Rear entry design
  • Expanded back to accommodate self-contained air breathing apparatus (SCBA)
  • Storm flap covers zipper which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper 
  • Vapor tight zipper is extra long to increase garment opening and aid donning and doffing
  • Attached internal gloves composed of five layer laminate film for a broad range of chemical protection
  • Neoprene gloves provide mechanical and physical durability to help protect inner gloves
  • Integrated socks composed of garment material with outer boot flaps
  • Double storm flap with hook and loop closure
  • Extra wide visor (3 layers) and attached dual layer gloves
  • Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
  • Two exhaust valves release higher pressure inside encapsulating garments while simultaneously helping to prevent inward leakage of external vapors or particles into garment
  • Valves include splash covers made of barrier material to help prevent liquid intrusion
  • Internal adjustable waist belt system for support and improved fit
  • Each suit has unique serial number and is fully tested at time of manufacture including positive air pressure integrity testing per ASTM F1052
Specifications
Specifications
Lime Yellow
3X-Large
Taped
Tychem
6 ft. 3 in. to 6 ft. 5 in.
31.5 in.