missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DuPont™ Tyvek™ 400 Disposable Pants

Pants with open ankle and elastic at waist for easy donning and removal . Lightweight, chemically and biologically inert, tear-resistant.

$305.72 - $441.87

Specifications

Color White
View More Specs

Products 10
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-067-055
View Documents
DuPont™
TY350SWHSM005000
Small
Case of 50 Each for $305.72
 
01-361-20B
View Documents
DuPont™
TY350SWHMD005000
Medium
Case of 50 Each for $329.88
 
19-067-054
View Documents
DuPont™
TY350SWHLG005000
Large
Case of 50 Each for $305.72
 
01-361-20E
View Documents
DuPont™
TY350SWH2X005000
2X-Large
Case of 50 Each for $349.72
 
01-361-20F
View Documents
DuPont™
TY350SWH3X005000
3X-Large
Case of 50 Each for $343.55
 
01-361-20D
View Documents
DuPont™
TY350SWHXL005000
X-Large
Case of 50 Each for $329.88
 
01-361-20G
View Documents
DuPont™
TY350SWH4X005000
4X-Large
Case of 50 Each for $370.97
 
17-500-293
View Documents
DuPont™
TY350SWH5X005000
5X-Large
Case of 50 Each for $393.24
 
17-500-294
View Documents
DuPont™
TY350SWH6X005000
6X-Large
Case of 50 Each for $416.83
 
17-500-295
View Documents
DuPont™
TY350SWH7X005000
7X-Large
Case of 50 Each for $441.87
 
Description

Description

 Tyvek™ garments are composed of flash spun high density polyethylene which creates a unique, nonwoven material. It provides an ideal balance of protection, durability and comfort of any limited use fabric technology.The fabric offers an inherent barrier against particles (down to 1.0 micron in size).
  • Suitable for applications that do not require full body coverage of coverall
  • Pants with elastic wrist, elastic waist and serged seams
  • Protection is built in fabric itself, there are no films or laminates to abrade or wear away
  • Fabric delivers consistently better barrier, even after wear and abrasion
  • Elasticized waist, No pockets, Open ankles, White
Applications

Lead and asbestos abatement/remedation, general maintenance/operations, spray painting, general clean-up.


Specifications

Specifications

White
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'