missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DuPont™ Tyvek™ IsoClean™ Boot Covers
Provides inherent barrier to dry particles, microorganisms, and non-hazardous liquids. Available in bulk or clean-processed packaging.
Supplier: DuPont™ IC457SWHLG01000B
Description
- Made from flashspun polyolefin protective material using proprietary flash spinning process
- Comfortable, lightweight and durable
- Ankle ties for secure fit
- Covered elastic opening
- Available with choice of seams, openings and soles
- Height: 18 in. (45.7cm)
- White
IC457S
- Serged seams with multiple interlocking threads sewn around raw edges of garment to create strong, stress-resistant seam
- With PVC soles that provide skid-resistance and durability
- Available bulk packaged in double transparent poly liners
- Bound seams covered with garment fabric to reinforce seam and to reduce potential for particle penetration
- With Gripper™ soles for enhanced skid-resistance and durability
- Available clean-processed
- Available bulk packaged in double transparent poly liners
Specifications
Large | |
Yes | |
Non-sterile | |
Anti-skid | |
5.4 mil | |
Bulk Packed | |
37.46 cm | |
46.99 cm |
Tyvek Flashspun Polyethylene | |
White | |
Elastic Top with Ankle Ties | |
Serged | |
16 CFR 1610 Class 1 | |
14.75 in. | |
18.5 in. | |
Class 1000 (ISO 6) |