missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DuPont™ Tyvek™ IsoClean™ Boot Covers
Provides inherent barrier to dry particles, microorganisms, and non-hazardous liquids. Available in bulk or clean-processed packaging.
$266.41 - $388.92
Specifications
Material | Tyvek Flashspun Polyethylene |
---|---|
Latex Free | Yes |
Color | White |
Sterility | Non-sterile |
Closure Type | Elastic Top with Ankle Ties |
Catalog Number | Mfr. No. | Size | Seam Type | Packaging Type | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Seam Type | Packaging Type | Price | Quantity | ||||
17-988-088
|
DuPont™
IC457SWHSM01000B |
Small | Serged | Bulk Packed |
Case of 100 Each for $266.41
|
|
||||
17-988-089
|
DuPont™
IC457SWHMD01000B |
Medium | Serged | Bulk Packed |
Case of 100 Each for $266.41
|
|
||||
17-988-090
|
DuPont™
IC457SWHLG01000B |
Large | Serged | Bulk Packed |
Case of 100 Each for $284.81
|
|
||||
17-988-091
|
DuPont™
IC457SWHXL01000B |
X-Large | Serged | Bulk Packed |
Case of 100 Each for $266.41
|
|
||||
17-988-100
|
DuPont™
IC458BWHMD01000B |
Medium | Bound | Bulk Packed |
Case of 100 Each for $290.88
|
|
||||
17-988-101
|
DuPont™
IC458BWHLG01000B |
Large | Bound | Bulk Packed |
Case of 100 Each for $320.12
|
|
||||
17-988-102
|
DuPont™
IC458BWHXL01000B |
X-Large | Bound | Bulk Packed |
Case of 100 Each for $320.12
|
|
||||
19-100-407
|
DuPont™
IC458BWHMD01000C |
Medium | Bound | Clean Processed |
Case of 100 Each for $377.81
|
|
||||
17-988-096
|
DuPont™
IC458BWHLG01000C |
Large | Bound | Clean Processed |
Case of 100 Each for $388.92
|
|
||||
17-988-097
|
DuPont™
IC458BWHXL01000C |
X-Large | Bound | Clean Processed |
Case of 100 Each for $377.81
|
|
||||
Description
- Made from flashspun polyolefin protective material using proprietary flash spinning process
- Comfortable, lightweight and durable
- Ankle ties for secure fit
- Covered elastic opening
- Available with choice of seams, openings and soles
- Height: 18 in. (45.7cm)
- White
IC457S
- Serged seams with multiple interlocking threads sewn around raw edges of garment to create strong, stress-resistant seam
- With PVC soles that provide skid-resistance and durability
- Available bulk packaged in double transparent poly liners
- Bound seams covered with garment fabric to reinforce seam and to reduce potential for particle penetration
- With Gripper™ soles for enhanced skid-resistance and durability
- Available clean-processed
- Available bulk packaged in double transparent poly liners
Specifications
Tyvek Flashspun Polyethylene | |
White | |
Elastic Top with Ankle Ties | |
16 CFR 1610 Class 1 | |
46.99 cm |
Yes | |
Non-sterile | |
Anti-skid | |
18.5 in. | |
Class 1000 (ISO 6) |