DuPont™ Tyvek™ IsoClean™ Serged Seam Cleanroom Frocks
Available in sterile or non-sterile versions, Tyvek™ cleanroom frocks provide an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash in ISO 5 (Class 100) and above environments.
Supplier:  DuPont™ IC262SWHXL00300S
 View more versions of this product
Catalog No. 17988064
Explore available promotions
Alert:
Garments made of Tyvek™ should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Description
Description
  • Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam.
  • Elastic opening for tighter fit at wrist
  • Mandarin collar with snap for secure fit
  • Body-length jacket design with front snap closures for easy donning and doffing
  • Raglan sleeve design for greater range of motion and comfort.
  • Generous sizing
  • Non-sterile versions are bulk packaged in double transparent poly liners
  • Sterile versions are individually packaged
  • Manufactured under specifications that do not contain natural rubber latex
  • Sterile frocks are gamma sterilized to SAL of 10-6
Specifications
Specifications
Controlled Environments Frock
Polyethylene
Single-use
Snaps
Serged
38 to 40 in.
Individually Packaged
121.28 cm
101.6 cm
X-Large
White
Sterile
Elastic
High Mandarin
Raglan Sleeve
47.75 in.
40 in.
Class 100 (ISO 5)