missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DuPont™ Tyvek™ IsoClean™ Serged Seam Cleanroom Frocks

Available in sterile or non-sterile versions, Tyvek™ cleanroom frocks provide an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash in ISO 5 (Class 100) and above environments.

$267.99 - $505.52

Specifications

Product Type Controlled Environments Frock
Color White
Closure Type Snaps
Seam Type Serged
Collar Type High Mandarin
View More Specs

Alert:

Garments made of Tyvek™ should not be used around heat, flames, sparks or in potentially flammable or explosive environments.


Products 11
Catalog Number Mfr. No. Size Sterility Packaging Type Price Quantity  
Catalog Number Mfr. No. Size Sterility Packaging Type Price Quantity  
17-988-072
View Documents
DuPont™
IC262SWH3X00300B
3X-Large Non-sterile Bulk Packaged
Case of 30 Each for $301.04
 
17988066
View Documents
DuPont™
IC262SWH3X00300S
3X-Large Sterile Individually Packaged
Case of 30 Each for $505.52
 
17988065
View Documents
DuPont™
IC262SWH2X00300S
2X-Large Sterile Individually Packaged
Case of 30 Each for $476.89
 
17-988-071
View Documents
DuPont™
IC262SWH2X00300B
2X-Large Non-sterile Bulk Packaged
Case of 30 Each for $284.01
 
17988070
View Documents
DuPont™
IC262SWHXL00300B
X-Large Non-sterile Bulk Packaged
Case of 30 Each for $267.99
 
17988064
View Documents
DuPont™
IC262SWHXL00300S
X-Large Sterile Individually Packaged
Case of 30 Each for $449.93
 
17-988-069
View Documents
DuPont™
IC262SWHLG00300B
Large Non-sterile Bulk Packaged
Case of 30 Each for $267.99
 
17-988-063
View Documents
DuPont™
IC262SWHLG00300S
Large Sterile Individually Packaged
Case of 30 Each for $449.93
 
17988068
View Documents
DuPont™
IC262SWHMD00300B
Medium Non-sterile Bulk Packaged
Case of 30 Each for $267.99
 
17-988-062
View Documents
DuPont™
IC262SWHMD00300S
Medium Sterile Individually Packaged
Case of 30 Each for $449.93
 
17-999-000
View Documents
DuPont™
IC262SWH4X00300B
4X-Large Non-sterile Bulk Packaged
Case of 30 Each for $319.09
 
Description

Description

  • Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam.
  • Elastic opening for tighter fit at wrist
  • Mandarin collar with snap for secure fit
  • Body-length jacket design with front snap closures for easy donning and doffing
  • Raglan sleeve design for greater range of motion and comfort.
  • Generous sizing
  • Non-sterile versions are bulk packaged in double transparent poly liners
  • Sterile versions are individually packaged
  • Manufactured under specifications that do not contain natural rubber latex
  • Sterile frocks are gamma sterilized to SAL of 10-6
Specifications

Specifications

Controlled Environments Frock
Snaps
High Mandarin
Class 100 (ISO 5)
White
Serged
Raglan Sleeve
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'