missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DuPont™ Tyvek™ IsoClean™ Serged Seam Cleanroom Frocks
Available in sterile or non-sterile versions, Tyvek™ cleanroom frocks provide an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash in ISO 5 (Class 100) and above environments.
Supplier: DuPont™ IC262SWH3X00300S
Alert:
Garments made of Tyvek™ should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Description
- Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam.
- Elastic opening for tighter fit at wrist
- Mandarin collar with snap for secure fit
- Body-length jacket design with front snap closures for easy donning and doffing
- Raglan sleeve design for greater range of motion and comfort.
- Generous sizing
- Non-sterile versions are bulk packaged in double transparent poly liners
- Sterile versions are individually packaged
- Manufactured under specifications that do not contain natural rubber latex
- Sterile frocks are gamma sterilized to SAL of 10-6
Specifications
Controlled Environments Frock | |
Polyethylene | |
Single-use | |
Snaps | |
Serged | |
43 to 46 in. | |
Individually Packaged | |
141.6 cm | |
115.57 cm |
3X-Large | |
White | |
Sterile | |
Elastic | |
High Mandarin | |
Raglan Sleeve | |
55.75 in. | |
45.5 in. | |
Class 100 (ISO 5) |