DuPont™ Tyvek™ IsoClean™ Serged Seam Cleanroom Frocks
Available in sterile or non-sterile versions, Tyvek™ cleanroom frocks provide an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash in ISO 5 (Class 100) and above environments.
Supplier:  DuPont™ IC262SWH3X00300B
 View more versions of this product
Catalog No. 17-988-072
Explore available promotions
Alert:
Garments made of Tyvek™ should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Description
Description
  • Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam.
  • Elastic opening for tighter fit at wrist
  • Mandarin collar with snap for secure fit
  • Body-length jacket design with front snap closures for easy donning and doffing
  • Raglan sleeve design for greater range of motion and comfort.
  • Generous sizing
  • Non-sterile versions are bulk packaged in double transparent poly liners
  • Sterile versions are individually packaged
  • Manufactured under specifications that do not contain natural rubber latex
  • Sterile frocks are gamma sterilized to SAL of 10-6
Specifications
Specifications
Controlled Environments Frock
Polyethylene
Single-use
Snaps
Serged
43 to 46 in.
Bulk Packaged
141.6 cm
115.57 cm
3X-Large
White
Non-sterile
Elastic
High Mandarin
Raglan Sleeve
55.75 in.
45.5 in.
Class 100 (ISO 5)