DuPont™ Tyvek™ IsoClean™ Sterile Sleeves
Tyvek sterile sleeves provides a barrier to particles, microorganisms and non-hazardous light liquid splash.
Supplier:  DuPont™ IC501BWH0001000S
 View more versions of this product
Catalog No. 17-988-109
Explore available promotions
Alert:
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Description
Description
Tyvek sterile sleeves provides a barrier to particles, microorganisms and non-hazardous light liquid splash.
  • Bound seams
  • Covered elastic wrist, bicep
  • Comfortable, lightweight and durable
  • Length: 18 in. (45.7cm)
  • White
Specifications
Specifications
Controlled Environment Sleeve
Tyvek
Elastic
ASTM F2101
45.7 cm
Standard
Single-use
IC501B
18 in.
Bulk Packaged