missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DuPont™ Tyvek™ IsoClean™ Sterile Sleeves
Tyvek sterile sleeves provides a barrier to particles, microorganisms and non-hazardous light liquid splash.
$366.23 - $370.68
Specifications
Size | Standard |
---|---|
Material | Tyvek |
Disposable | Single-use |
Cuff Style | Elastic |
Model | IC501B |
Alert:
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Catalog Number | Mfr. No. | Packaging | Thickness | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Packaging | Thickness | Price | Quantity | ||||
17-988-110
|
DuPont™
IC501BWH000100CS |
Clean Processed | 5.0 mil |
Case of 100 Pair for $370.68
|
|
||||
17-988-109
|
DuPont™
IC501BWH0001000S |
Bulk Packaged |
Case of 100 Each for $366.23
|
|
|||||
Description
Tyvek sterile sleeves provides a barrier to particles, microorganisms and non-hazardous light liquid splash.
- Bound seams
- Covered elastic wrist, bicep
- Comfortable, lightweight and durable
- Length: 18 in. (45.7cm)
- White
Specifications
Standard | |
Single-use | |
IC501B | |
18 in. |
Tyvek | |
Elastic | |
ASTM F2101 | |
45.7 cm |