DuPont™ Tyvek™ 400 Shirts
Lightweight, chemically and biologically inert, tear-resistant
Supplier:  DuPont™ TY303SWHSM005000
 View more versions of this product
Catalog No. 01-361-18A
Explore available promotions
Alert:
Garments made of Tyvek should not be used around heat, flames, sparks, or in potentially flammable or explosive environments.
Description
Description
Inherent barrier protection. Stops submicron particles, even after abrasion.
Shirts
  • Snap front
  • Full-length sleeves with open cuffs
  • White
Specifications
Specifications
Small
White
Snaps
Lead and asbestos abatement/remedation, general maintenance/operations, spray painting, general clean-up
77.47 cm
Tyvek
Serged
Shirt
30.5 in.