Fisherbrand™ Durac™ Plus™ Thermometers
Ideal for use in applications in which a very precise, accurate temperature measurement is required.
$25.51 - $46.15
Specifications
For Use With (Application) Chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories
Material Glass
Thermometer Fill Blue Spirit
View More Specs
Products 4
Catalog Number Mfr. No. Temperature Range (Metric) Immersion Type Length (Metric) Overall Subdivision Price Quantity  
Catalog Number Mfr. No. Temperature Range (Metric) Immersion Type Length (Metric) Overall Subdivision Price Quantity  
S02004G
View Documents
Fisherbrand™
B620070422
-10°C to +260°C Partial Immersion 300 mm 2°C
Each for $46.15
Case of 25 Each for $1,153.75
S02004K
View Documents
Fisherbrand™
B620080122
-20°C to +110°C Total Immersion 300 mm 1°C
Each for $25.51
Case of 25 Each for $637.75
S02004D
View Documents
Fisherbrand™
B620070322
-20°C to +150°C Partial Immersion 300 mm 1°C
Each for $26.26
Case of 25 Each for $656.50
S02004M
View Documents
Fisherbrand™
B620080322
-10°C to +225°C Total Immersion 300 mm 2°C
Each for $41.36
Case of 25 Each for $1,034.00
Description
Description
  • Triangular anti-roll fitting
  • Permanently fused markings
  • Blue spirit liquid with white background glass
  • Individually packaged
  • Individually serialized with a traceability statement
Certifications
Tested and calibrated in a triple accredited/registered ISO/IEC 17025:2005, A2LA™ Laboratory.
Specifications
Specifications
Chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories
Glass
Blue Spirit
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'