Fisherbrand™ Durac™ Plus™ Thermometers
Ideal for use in applications in which a very precise, accurate temperature measurement is required.
Supplier: Fisherbrand™ B620080322
Description
- Triangular anti-roll fitting
- Permanently fused markings
- Blue spirit liquid with white background glass
- Individually packaged
- Individually serialized with a traceability statement
Certifications
Tested and calibrated in a triple accredited/registered ISO/IEC 17025:2005, A2LA™ Laboratory.
Specifications
Chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories | |
12 in. | |
Glass | |
225°C | |
-10°C to +225°C |
Total Immersion | |
300 mm | |
Blue Spirit | |
-10°C | |
2°C |