DWK Life Sciences DURAN™ HPLC Bottle with DIN Thread, GL 45
With easy-to-read scale and large labelling field for easy marking, in fired-on, highly durable white ceramic.
Supplier:  DWK Life Sciences 1129821
 View more versions of this product
Catalog No. 09-841-007
Explore available promotions
Description
Description
Complete system comprising: DURAN™ pressure plus laboratory bottle with 4-port screw cap (PP); four screw connections (black, M8 thread) and silicone seals. Connection of different hose diameters (1.6 mm and 3.2 mm) as well as sterile pressure equalisation sets (membrane filter 0.2 ++m) is possible. Unused ports can be sealed with silicone blanking seals.
Safe transfer of liquid media within a closed and sterile system (evaporation is reduced).
Specifications
Specifications
16 oz., 500mL
16 oz.
Glass
7.0 in.
500 mL
3.38 in.
176 mm
86 mm