DWK Life Sciences DURAN™ HPLC Bottle with DIN Thread, GL 45
With easy-to-read scale and large labelling field for easy marking, in fired-on, highly durable white ceramic.
$682.74 - $865.24
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
09-841-007
|
DWK Life Sciences
1129821 |
500 mL |
Case of 2 Each for $682.74
|
|
|||||
09-841-008
|
DWK Life Sciences
1129820 |
1000 mL |
Case of 2 Each for $865.24
|
|
|||||
Description
Complete system comprising: DURAN™ pressure plus laboratory bottle with 4-port screw cap (PP); four screw connections (black, M8 thread) and silicone seals. Connection of different hose diameters (1.6 mm and 3.2 mm) as well as sterile pressure equalisation sets (membrane filter 0.2 ++m) is possible. Unused ports can be sealed with silicone blanking seals.
Safe transfer of liquid media within a closed and sterile system (evaporation is reduced).
Specifications
Glass |