DWK Life Sciences DURAN™ HPLC Bottle with DIN Thread, GL 45
With easy-to-read scale and large labelling field for easy marking, in fired-on, highly durable white ceramic.
Supplier:  DWK Life Sciences 1129820
 View more versions of this product
Catalog No. 09-841-008
Explore available promotions
Description
Description
Complete system comprising: DURAN™ pressure plus laboratory bottle with 4-port screw cap (PP); four screw connections (black, M8 thread) and silicone seals. Connection of different hose diameters (1.6 mm and 3.2 mm) as well as sterile pressure equalisation sets (membrane filter 0.2 ++m) is possible. Unused ports can be sealed with silicone blanking seals.
Safe transfer of liquid media within a closed and sterile system (evaporation is reduced).
Specifications
Specifications
32 oz., 1000mL
32 oz.
Glass
8.85 in.
1000 mL
3.9 in.
225 mm
101 mm