Eagle™ Galvanized-Steel Safety Cans
For safe storage and handling of gasoline and other flammable liquids
Supplier:  Eagle™ UI50S
 View more versions of this product
Catalog No. 04-664C
Explore available promotions
Description
Description
Seamless, 24-gauge galvanized steel body for durability and corrosion resistance.
  • Spring-closing lid with a neoprene gasket safely relieves internal pressure at 5psig
  • Brass flame arrestor in spout allows for safe filling and dispensing of flammable liquids and provides flashback protection
  • Metal spout pours accurately, prevents static sparks
  • Finished in bright red, yellow (for diesel), or blue (for kerosene; contact your Fisher Safety Customer Service Representative to order) enamel paint
  • Yellow silk-screened label in English, Spanish, and French
  • Lead free
Certifications
FM, UL, and ULC approved.
Compliance
Meet OSHA and NFPA Code 30 requirements.
Specifications
Specifications
Safety Can
5 gal.
Red
Squeeze handle, pour spout, flame arrestor, Kerosene printed on can
342 mm
317 mm
FM, UL, ULC, OSHA 29 CFR 1910.106, NFPA 30
18.9 L
Galvanized Steel
13.5 in.
12.5 in.