Eagle™ Galvanized-Steel Safety Cans
For safe storage and handling of gasoline and other flammable liquids
$122.17 - $136.04
Specifications
Product Type Safety Can
Color Red
View More Specs
Products 2
Catalog Number Mfr. No. Material Product Type Height (Metric) Price Quantity  
Catalog Number Mfr. No. Material Product Type Height (Metric) Price Quantity  
04-664A
View Documents
Eagle™
UI10S
Nonterneplate Steel Safety Can 203 mm
Each for $122.17
04-664C
View Documents
Eagle™
UI50S
Galvanized Steel Safety Can 342 mm
Each for $136.04
Description
Description
Seamless, 24-gauge galvanized steel body for durability and corrosion resistance.
  • Spring-closing lid with a neoprene gasket safely relieves internal pressure at 5psig
  • Brass flame arrestor in spout allows for safe filling and dispensing of flammable liquids and provides flashback protection
  • Metal spout pours accurately, prevents static sparks
  • Finished in bright red, yellow (for diesel), or blue (for kerosene; contact your Fisher Safety Customer Service Representative to order) enamel paint
  • Yellow silk-screened label in English, Spanish, and French
  • Lead free
Certifications
FM, UL, and ULC approved.
Compliance
Meet OSHA and NFPA Code 30 requirements.
Specifications
Specifications
Safety Can
Red
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'