missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Eagle™ Polyethylene Oily Waste Cans
Designed for temporary storage of solvent, waste rags, and other materials subject to spontaneous combustion
Supplier: Eagle™ 933FLY
Description
Durable, fire-safe cans won't rust or dent and resist most chemical compounds.
- Cans feature foot treadle that opens cover; self-closing lid
- Choice of red or high-visibility yellow
Certifications
FM approved.
Compliance
Meet OSHA 29CFR1910.108 and 20CFR1910.106 requirements.
Specifications
23 L | |
13.5 in. | |
34.29 cm | |
Yellow | |
1 Ea. | |
4 kg | |
Foot lever | |
16.5 in. | |
HDPE |
FM, OSHA 29 CFR 1910.108, 20 CFR 1910.106 | |
Self Closing | |
6 gal. | |
41 cm | |
9 lb. | |
41.6 cm | |
For temporary storage of solvent, waste rags, and other materials subject to spontaneous combustion | |
16 in. | |
Oily Waste Can |