missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Eagle™ Step-On Biohazard Waste Containers
Provide safe, temporary collection of biological waste (no sharps). Foot operated pedal with self-closing lid reduces the risk of contamination. Alert workers to potential biohazards and meet OSHA requirements for exposures to bloodborne pathogens.
Supplier: Eagle™ 943BIO
Description
- Suitable for medical facilities and research labs
- Safely collect biological waste; reduce contamination
- Foot lever opens container for hands-free operation
- Heavy-wall, high density polyethylene construction won’t rust or dent
- Labeled to advise workers of potential biohazards
Order Info
Use with Fisherbrand™ Biohazard Autoclavable Bags; sold separately.
Certifications
FM approved.
Compliance
Comply with 29 CFR 1910.1030 requirements for exposure to bloodborne pathogens.
Specifications
22.7 L | |
41 cm | |
6 gal. | |
16.5 x 13.5 x 16 in. (42 x 34 x 41 cm) | |
42 cm | |
Prominent biohazard warning label, foot lever | |
13.5 in. | |
16 in. | |
Biohazard Waste Can |
FM, OSHA 29 CFR 1910.1030, NFPA | |
16 in. | |
Red | |
41 cm | |
34 cm | |
Suitable for medical facilities and research labs | |
16.5 in. | |
HDPE |