missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eagle™ Step-On Biohazard Waste Containers

Provide safe, temporary collection of biological waste (no sharps). Foot operated pedal with self-closing lid reduces the risk of contamination. Alert workers to potential biohazards and meet OSHA requirements for exposures to bloodborne pathogens.

$177.49 - $242.99

Specifications

Certifications/Compliance FM, OSHA 29 CFR 1910.1030, NFPA
Color Red
Includes Prominent biohazard warning label, foot lever
For Use With (Application) Suitable for medical facilities and research labs
Material HDPE
View More Specs

Products 3
Catalog Number Mfr. No. Capacity (English) Height (Metric) Length (Metric) Width (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (English) Height (Metric) Length (Metric) Width (Metric) Price Quantity  
18-880A
View Documents
Eagle™
943BIO
6 gal. 41 cm 42 cm 34 cm
Each for $177.49
 
18-880B
View Documents
Eagle™
945BIO
10 gal. 46 cm 56 cm 45.7 cm
Each for $237.93
 
18-880C
View Documents
Eagle™
947BIO
14 gal. 53 cm 56 cm 45.7 cm
Each for $242.99
 
Description

Description

 

  • Suitable for medical facilities and research labs
  • Safely collect biological waste; reduce contamination
  • Foot lever opens container for hands-free operation
  • Heavy-wall, high density polyethylene construction won’t rust or dent
  • Labeled to advise workers of potential biohazards

Order Info

Use with Fisherbrand™ Biohazard Autoclavable Bags; sold separately.

Certifications

FM approved.

Compliance

Comply with 29 CFR 1910.1030 requirements for exposure to bloodborne pathogens.

Specifications

Specifications

FM, OSHA 29 CFR 1910.1030, NFPA
Prominent biohazard warning label, foot lever
HDPE
Red
Suitable for medical facilities and research labs
Biohazard Waste Can
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'