LevGo Eco-smartFunnel™ Disposable Eco Friendly Paper Lab Funnel
Eco-smartFunnel is a simple, cost-effective solution to reduce plastic use in the lab and reduce cross contamination from reuse of washed funnels
Supplier:  LevGo 19630
 View more versions of this product
Catalog No. 18-001-023
Explore available promotions
Description
Description
  • Reduce cross contamination
  • Fold flat design uses less storage space
  • Great for powders and cold liquids
  • Can be used in Saliva Collection Applications
  • Eco friendly, biodegradable
  • Made in the USA
Specifications
Specifications
5 x 2.25 in. (12.7 x 5.7 mm)
5 g
Disposable Eco Funnel
Sample collection, pouring viscous liquids and powders
Square
Single-use
Paper Disposable Funnel
Yes
FSC Certified Food grade clay coated paperboard