missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Elaidic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 117730010

 View more versions of this product

Catalog No. AC117730010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Elaidic acid
43.0°C to 45.0°C
>110°C
Glass bottle
CH3(CH2)7CH=CH(CH2)7CO2H
02,II,441
elaidic acid, trans-oleic acid, trans-9-octadecenoic acid, 9-octadecenoic acid, e-oleic acid, 9-octadecenoic acid, e, e-octadec-9-enoic acid, trans-elaidic acid, 9-octadecenoic acid, 9e, elaidinsaure
CCCCCCCCC=CCCCCCCCC(=O)O
282.46
CHEBI:27997
98%
112-79-8
288.0°C (100.0 mmHg)
97.5% min. (GC)
C18H34O2
1 g
15,3581
ZQPPMHVWECSIRJ-MDZDMXLPSA-N
(E)-octadec-9-enoic acid
637517
282.46

Chemical Identifiers

112-79-8
282.46
elaidic acid, trans-oleic acid, trans-9-octadecenoic acid, 9-octadecenoic acid, e-oleic acid, 9-octadecenoic acid, e, e-octadec-9-enoic acid, trans-elaidic acid, 9-octadecenoic acid, 9e, elaidinsaure
CHEBI:27997
CCCCCCCCC=CCCCCCCCC(=O)O
C18H34O2
ZQPPMHVWECSIRJ-MDZDMXLPSA-N
637517
(E)-octadec-9-enoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Warning

EINECSNumber : 204-006-6

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only