missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Elaidic acid, 98%, Thermo Scientific Chemicals

$195.87 - $195.87

Chemical Identifiers

CAS 112-79-8
Molecular Formula C18H34O2
Molecular Weight (g/mol) 282.46
InChI Key ZQPPMHVWECSIRJ-MDZDMXLPSA-N
Synonym elaidic acid, trans-oleic acid, trans-9-octadecenoic acid, 9-octadecenoic acid, e-oleic acid, 9-octadecenoic acid, e, e-octadec-9-enoic acid, trans-elaidic acid, 9-octadecenoic acid, 9e, elaidinsaure
PubChem CID 637517
ChEBI CHEBI:27997
IUPAC Name (E)-octadec-9-enoic acid
SMILES CCCCCCCCC=CCCCCCCCC(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC117730010
View Documents
Thermo Scientific Chemicals
117730010
1 g Glass bottle
Each for $195.87
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

112-79-8
282.46
elaidic acid, trans-oleic acid, trans-9-octadecenoic acid, 9-octadecenoic acid, e-oleic acid, 9-octadecenoic acid, e, e-octadec-9-enoic acid, trans-elaidic acid, 9-octadecenoic acid, 9e, elaidinsaure
CHEBI:27997
CCCCCCCCC=CCCCCCCCC(=O)O
C18H34O2
ZQPPMHVWECSIRJ-MDZDMXLPSA-N
637517
(E)-octadec-9-enoic acid

Specifications

112-79-8
288.0°C (100.0 mmHg)
97.5% min. (GC)
C18H34O2
1 g
15,3581
ZQPPMHVWECSIRJ-MDZDMXLPSA-N
(E)-octadec-9-enoic acid
637517
282.46
Elaidic acid
43.0°C to 45.0°C
>110°C
Glass bottle
CH3(CH2)7CH=CH(CH2)7CO2H
02,II,441
elaidic acid, trans-oleic acid, trans-9-octadecenoic acid, 9-octadecenoic acid, e-oleic acid, 9-octadecenoic acid, e, e-octadec-9-enoic acid, trans-elaidic acid, 9-octadecenoic acid, 9e, elaidinsaure
CCCCCCCCC=CCCCCCCCC(=O)O
282.46
CHEBI:27997
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Warning

EINECSNumber : 204-006-6

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only