MilliporeSigma™ Accessories for Pellicon XL Devices
For use with Pellicon XL Device
Supplier:  MilliporeSigma™ XXRES05ST
 View more versions of this product
Catalog No. XXRES05ST
Explore available promotions
Description
Description
  • 100mL Reservoir and Stand: Used with the Pellicon XL Device for simple separations of small volumes; stand mounts reservoir on Labscale™ system
  • Analog Pressure Gauge Kit (0-60psi): can be used with the Pellicon XL Device for accurate scaling studies; includes one pressure gauge and connection fittings
  • Multimanifold: Mounts directly onto either Labscale system reservoir or standard peristaltic pump to allow simultaneous use of three Pellicon XL Devices
  • Pellicon XL Device Stand: Supports one Pellicon XL Device
Specifications
Specifications
Pellicon XL 50 Cassette, Labscale TFF System
Simple separations of small volumes, for mounting 100mL reservoir on the Labscale™ system
1 Pack
Reservoir Stand