MilliporeSigma™ Accessories for Pellicon XL Devices
For use with Pellicon XL Device
$346.20 - $716.79
Specifications
For Use With (Equipment) | Pellicon XL 50 Cassette, Labscale TFF System |
---|
Catalog Number | Mfr. No. | Type | Quantity | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Quantity | Price | Quantity | ||||
XX42RES05
|
MilliporeSigma™
XX42RES05 |
Reservoir | 1 Pack |
N/A
|
|
||||
XXPXLGAGE
|
MilliporeSigma™
XXPXLGAGE |
Analog Pressure Gauge Kit | 1 Kit |
N/A
|
|
||||
XXRES05ST
|
MilliporeSigma™
XXRES05ST |
Reservoir Stand | 1 Pack |
N/A
|
|
||||
XXPXLSTND
|
MilliporeSigma™
XXPXLSTND |
Pellicon XL Stand | 1 Pack |
N/A
|
|
||||
Description
- 100mL Reservoir and Stand: Used with the Pellicon XL Device for simple separations of small volumes; stand mounts reservoir on Labscale™ system
- Analog Pressure Gauge Kit (0-60psi): can be used with the Pellicon XL Device for accurate scaling studies; includes one pressure gauge and connection fittings
- Multimanifold: Mounts directly onto either Labscale system reservoir or standard peristaltic pump to allow simultaneous use of three Pellicon XL Devices
- Pellicon XL Device Stand: Supports one Pellicon XL Device
Specifications
Pellicon XL 50 Cassette, Labscale TFF System |