missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ Immobilon™ Block Noise Canceling Reagents
Minimize background in Western blots, increasing sensitivity for superior detection of even low-abundance proteins
Supplier: MilliporeSigma™ WBAVDFL01
Description
- Ready-to-use
- Protein-free, reducing nonspecific binding, especially when using phosphoantibodies or avidin-biotin systems
- Specially engineered for the EMD Millipore SNAP i.d.™ detection system, but may be used in traditional Western blotting protocols
- Suitable for the dilution of primary and secondary antibodies
- Sufficient for 19 miniblots (each 8 × 7cm blot requires at least 26mL for SNAP i.d.)
- Compatible with streptavidin antibodies
- Compatible with PVDF (Immobilon™ P, Immobilon FL) and nitrocellulose
- Block-CH reagent is compatible with Immobilon HRP substrate
- Choose from three Block reagents
- Block-CH reagent for HRP chemiluminescence detection
- Block-FL reagent for fluorescence detection
- Block-PO reagent for phosphoprotein detection via chemiluminescence or fluorescence
Specifications
Block-FL Reagent | |
500 mL | |
20°C to 25°C |
Fluorescence Assay | |
Nineteen 8 x 7cm miniblots |