MilliporeSigma™ HPTLC RP-modified Silica Gel 60 Plates, Glass Backed
Glass HPTLC Silica gel 60 RP plates
Supplier: MilliporeSigma™ 1137240001
Description
RP modified plates serve two purposes: act as a pilot method for HPLC and allow to choose various solvent system for special separations. Our RP-modified silica layers are well suited for many separation challenges that unmodified silica cannot overcome. These layers use aqueous solvent systems to separate extremely non-polar substances and analyze particular polar substances that can adapt to ion-pair chromatography. Whatâs more, RP-modified silica layers are less dependent on atmospheric humidity. Unlike unmodified silica, RP-phases do not exhibit catalytic activity. This makes them the plates of choice for unstable substance that tend to experience oxidative degradation.Specifications
Glass | |
60 Å | |
RP-modified Silica HPTLC Plate | |
Separation of Extremely Non-Polar and Highly Polar Substances Using Aqueous Solvent Systems, Fast Analysis of Complex Samples for Manual or Instrumental Use | |
200 μm | |
10 cm |
Glass | |
25/Pk. | |
254 nm | |
Silica Gel 60 RP-18 F254 s | |
10 x 10 cm | |
10 cm |