MilliporeSigma™ HPTLC RP-modified Silica Gel 60 Plates, Glass Backed
Glass HPTLC Silica gel 60 RP plates
$1407.24 - $2615.21
Specifications
Backing Material | Glass |
---|---|
Material | Glass |
Pore Size | 60 Å |
For Use With (Application) | Separation of Extremely Non-Polar and Highly Polar Substances Using Aqueous Solvent Systems, Fast Analysis of Complex Samples for Manual or Instrumental Use |
Thickness (Metric) | 200 μm |
Catalog Number | Mfr. No. | Dimensions (L x W) | Quantity | Sorbent | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Quantity | Sorbent | Price | Quantity | |||
M1059140001
|
MilliporeSigma™
1059140001 |
20 x 10 cm | 25/Pk. | Silica Gel 60 RP-18 |
Each for $1,684.37
|
|
|||
M1131240001
|
MilliporeSigma™
1131240001 |
10 x 10 cm | 25/Pk. | Silica Gel 60 RP-18 W F254 s |
Each for $1,407.24
|
|
|||
M1137240001
|
MilliporeSigma™
1137240001 |
10 x 10 cm | 25/Pk. | Silica Gel 60 RP-18 F254 s |
Each for $2,009.24
|
|
|||
M1137250001
|
MilliporeSigma™
1137250001 |
10 x 10 cm | 25/Pk. | Silica Gel 60 RP-8 F254 s |
Each for $1,828.61
|
|
|||
M1137260001
|
MilliporeSigma™
1137260001 |
10 x 10 cm | 25/Pk. | Silica Gel 60 RP-2 F254 s |
Each for $2,009.24
|
|
|||
M1142960001
|
MilliporeSigma™
1142960001 |
20 x 10 cm | 25/Pk. | Wettable Silica Gel RP-18 W |
Each for $2,615.21
|
|
|||
M1162250001
|
MilliporeSigma™
1162250001 |
10 x 20 cm | 50/Pk. | Silica Gel 60 RP-18 Modified |
Each for $1,468.34
|
|
|||
Description
RP modified plates serve two purposes: act as a pilot method for HPLC and allow to choose various solvent system for special separations. Our RP-modified silica layers are well suited for many separation challenges that unmodified silica cannot overcome. These layers use aqueous solvent systems to separate extremely non-polar substances and analyze particular polar substances that can adapt to ion-pair chromatography. Whatâs more, RP-modified silica layers are less dependent on atmospheric humidity. Unlike unmodified silica, RP-phases do not exhibit catalytic activity. This makes them the plates of choice for unstable substance that tend to experience oxidative degradation.Specifications
Glass | |
60 Å | |
200 μm |
Glass | |
Separation of Extremely Non-Polar and Highly Polar Substances Using Aqueous Solvent Systems, Fast Analysis of Complex Samples for Manual or Instrumental Use | |
10 cm |