MilliporeSigma™ HPTLC RP-modified Silica Gel 60 Plates, Glass Backed
Glass HPTLC Silica gel 60 RP plates
Supplier:  MilliporeSigma™ 1137260001
 View more versions of this product
Catalog No. M1137260001
Explore available promotions
Description
Description
RP modified plates serve two purposes: act as a pilot method for HPLC and allow to choose various solvent system for special separations. Our RP-modified silica layers are well suited for many separation challenges that unmodified silica cannot overcome. These layers use aqueous solvent systems to separate extremely non-polar substances and analyze particular polar substances that can adapt to ion-pair chromatography. What’s more, RP-modified silica layers are less dependent on atmospheric humidity. Unlike unmodified silica, RP-phases do not exhibit catalytic activity. This makes them the plates of choice for unstable substance that tend to experience oxidative degradation.
Specifications
Specifications
Glass
60 Å
RP-modified Silica HPTLC Plate
Separation of Extremely Non-Polar and Highly Polar Substances Using Aqueous Solvent Systems, Fast Analysis of Complex Samples for Manual or Instrumental Use
200 μm
10 cm
Glass
25/Pk.
254 nm
Silica Gel 60 RP-2 F254 s
10 x 10 cm
10 cm