MilliporeSigma™ MultiScreen™ 96-Well Assay Plates for ELISPOT Assays
Give optimal results in immunological ELISPOT (enzyme-linked immunospot) assays by providing high protein-binding capacity with low background staining
Supplier: MilliporeSigma™ MAIPSWU10
Description
Give optimal results in immunological ELISPOT (enzyme-linked immunospot) assays by providing high protein-binding capacity with low background staining
- Membrane-to-plate seal ensures no reagent crosstalk and limited optical crosstalk Patented underdrain support structure allows liquid volumes to be maintained for incubation times of up to 21 days
Specifications
Standard | |
MultiScreen™HTS plate with Immobilon™-P membrane without underdrain | |
0.45 μm | |
Acrylic | |
Sterile | |
ELISPOT assays |
Hydrophobic PVDF | |
8 x 12 | |
White | |
96 | |
0.28cm2 |