missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MilliporeSigma™ MultiScreen™ 96-Well Assay Plates for ELISPOT Assays

Give optimal results in immunological ELISPOT (enzyme-linked immunospot) assays by providing high protein-binding capacity with low background staining

$461.00 - $1911.34

Specifications

Format Standard
Array 8 x 12
Pore Size 0.45 μm
No. of Wells 96
For Use With (Application) ELISPOT assays
View More Specs

Products 6
Catalog Number Mfr. No. Description Color Material Sterility Price Quantity  
Catalog Number Mfr. No. Description Color Material Sterility Price Quantity  
MAHAS4510
View Documents
MilliporeSigma™
MAHAS4510
MultiScreen™-HA with MCE membrane Clear Styrene Sterile
Pack of 10 Each for $483.26
 
MAIPS4510
View Documents
MilliporeSigma™
MAIPS4510
MultiScreen™-IP plate with Immobilon™-P membrane without underdrain Clear ABS Plastic Sterile
Pack of 10 Each for $461.00
 
MSIPN4W50
View Documents
MilliporeSigma™
MSIPN4W50
MultiScreen-IP Filter Plate with Immobilon™-P membrane White ABS Plastic Non-sterile
Pack of 50 Each for $1,911.34
 
MSIPS4W10
View Documents
MilliporeSigma™
MSIPS4W10
MultiScreen™HTS-IP plate with Immobilon™-P membrane White Acrylic Sterile
Pack of 10 Each for $513.46
 
S2EM004M99
View Documents
MilliporeSigma™
S2EM004M99
MultiScreen-IP Filter Plate with Immobilon™-P membrane White Acrylic Sterile
Pack of 10 Each for $605.28
 
35-1044
View Documents
MilliporeSigma™
MAIPSWU10
MultiScreen™HTS plate with Immobilon™-P membrane without underdrain White Acrylic Sterile
Pack of 10 Each for $628.00
 
Description

Description

Give optimal results in immunological ELISPOT (enzyme-linked immunospot) assays by providing high protein-binding capacity with low background staining

  • Membrane-to-plate seal ensures no reagent crosstalk and limited optical crosstalk Patented underdrain support structure allows liquid volumes to be maintained for incubation times of up to 21 days

Specifications

Specifications

Standard
0.45 μm
ELISPOT assays
8 x 12
96
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'