MilliporeSigma™ MultiScreen™ 96-Well Assay Plates for ELISPOT Assays
Give optimal results in immunological ELISPOT (enzyme-linked immunospot) assays by providing high protein-binding capacity with low background staining
Supplier: MilliporeSigma™ MAIPS4510
Description
Give optimal results in immunological ELISPOT (enzyme-linked immunospot) assays by providing high protein-binding capacity with low background staining
- Membrane-to-plate seal ensures no reagent crosstalk and limited optical crosstalk Patented underdrain support structure allows liquid volumes to be maintained for incubation times of up to 21 days
Specifications
Standard | |
MultiScreen™-IP plate with Immobilon™-P membrane without underdrain | |
0.45 μm | |
ABS Plastic | |
Sterile | |
ELISPOT assays |
Hydrophobic PVDF | |
8 x 12 | |
Clear | |
96 | |
0.28cm2 | |
10/Pk. |